Order Entry
Denmark
Orders LinkContactUsLinkComponent
6-Bromquinolin

6-Bromquinolin

Bottle, 10 g
Katalog # APOSOR23076-10G
Ætsende
Irriterende
Leverandør:   Apollo Scientific
6-Bromquinolin
Katalog # APOSOR23076-10G
Leverandør:  Apollo Scientific
CAS nummer:  5332-25-2
6-Bromquinolin

Bottle, 10 g

CAS nummer:  5332-25-2
Price
165,00 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents
  • CAS nummer:5332-25-2
  • Formel:C₉H₆BrN
  • MDL nummer:MFCD00024023
  • Molecular Weight:208.06
  • Smeltende Pt:24 °C
  • Tæthed:1,538 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    10 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.