Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2,4-Pentandion

2,4-Pentandion

Bottle, 500 mL
Katalog # APOSOR16430-500ML
Brandfarlig
Giftig
Leverandør:   Apollo Scientific
2,4-Pentandion
Katalog # APOSOR16430-500ML
Leverandør:  Apollo Scientific
CAS nummer:  123-54-6
2,4-Pentandion

Bottle, 500 mL

CAS nummer:  123-54-6
Price
593,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Synonymer:  ACAC, 2,4-Pentanedione
  • CAS nummer:123-54-6
  • Formel:CH₃COCH₂COCH₃
  • MDL nummer:MFCD00008787
  • Molecular Weight:100.12 g/mol
  • Kogende Pt:140…141 °C
  • Smeltende Pt:–23 °C
  • Tæthed:0,975 g/cm³ (25 °C)
  • Flash Pt:38 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-634-0
  • FN:2310
  • ADR:3+6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    500 mL

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.