Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2,4-Pentandion

2,4-Pentandion

Bottle, 2,5 L
Katalog # APOSOR16430-2.5L
Brandfarlig
Giftig
Leverandør:   Apollo Scientific
2,4-Pentandion
Katalog # APOSOR16430-2.5L
Leverandør:  Apollo Scientific
CAS nummer:  123-54-6
2,4-Pentandion

Bottle, 2,5 L

CAS nummer:  123-54-6
Price
1.890,00 kr
Log ind for tilgængelighed
1 * 2,5 L
Product Details & Documents
Synonymer:  ACAC, 2,4-Pentanedione
  • CAS nummer:123-54-6
  • Formel:CH₃COCH₂COCH₃
  • MDL nummer:MFCD00008787
  • Molecular Weight:100.12 g/mol
  • Kogende Pt:140…141 °C
  • Smeltende Pt:–23 °C
  • Tæthed:0,975 g/cm³ (25 °C)
  • Flash Pt:38 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-634-0
  • FN:2310
  • ADR:3+6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    2,5 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.