Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-Aminopyrazin

2-Aminopyrazin

Bottle, 100 g
Katalog # APOSOR5470-100G
Irriterende
Leverandør:   Apollo Scientific
2-Aminopyrazin
Katalog # APOSOR5470-100G
Leverandør:  Apollo Scientific
CAS nummer:  5049-61-6
2-Aminopyrazin

Bottle, 100 g

CAS nummer:  5049-61-6
Price
363,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  Aminopyrazine, Pyrazin-2-amine
  • CAS nummer:5049-61-6
  • Formel:C₄H₅N₃
  • MDL nummer:MFCD00006137
  • Molecular Weight:95.1 g/mol
  • Smeltende Pt:118…121 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:225-748-7
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.