Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
3-Hydroxybenzonitril

3-Hydroxybenzonitril

Bottle, 5 g
Katalog # APOSOR6704-5G
Irriterende
Leverandør:   Apollo Scientific
3-Hydroxybenzonitril
Katalog # APOSOR6704-5G
Leverandør:  Apollo Scientific
CAS nummer:  873-62-1
3-Hydroxybenzonitril

Bottle, 5 g

CAS nummer:  873-62-1
Price
187,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol
  • CAS nummer:873-62-1
  • Formel:C₇H₅NO
  • MDL nummer:MFCD00002252
  • Molecular Weight:119.12 g/mol
  • Smeltende Pt:78…81 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:212-845-4
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    5 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.