Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
3-Hydroxybenzonitril

3-Hydroxybenzonitril

Bottle, 25 g
Katalog # APOSOR6704-25G
Irriterende
Leverandør:   Apollo Scientific
3-Hydroxybenzonitril
Katalog # APOSOR6704-25G
Leverandør:  Apollo Scientific
CAS nummer:  873-62-1
3-Hydroxybenzonitril

Bottle, 25 g

CAS nummer:  873-62-1
Price
341,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol
  • CAS nummer:873-62-1
  • Formel:C₇H₅NO
  • MDL nummer:MFCD00002252
  • Molecular Weight:119.12 g/mol
  • Smeltende Pt:78…81 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:212-845-4
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.