Order Entry
Denmark
Orders LinkContactUsLinkComponent
2'-Fluor-4'-methoxyacetophenon 98%
Bottle, 5 g
  APOSPC3817B-5G
2'-Fluor-4'-methoxyacetophenon 98%
  APOSPC3817B-5G
 :  74457-86-6
2'-Fluor-4'-methoxyacetophenon 98%
Bottle, 5 g
 :  74457-86-6
kr165.00
1 * 5 g
Precursor to phenethylthiazolythiourea (PETT) derivatives as potential HIV-1 reverse transcriptase inhibitors see: J. Med. Chem., 39, 4261, (1996).
  •  :74457-86-6
  •  :C₉H₉FO₂
  •  :MFCD00042290
  •  :168.17
  •  :80…85 °C/0,1mmHg
  •  :51…53 °C
  •  :>110 °C
  •  :Stue temperatur
  • Pakning:
    Bottle
  • Pakn:
    5 g