2'-Fluor-4'-methoxyacetophenon 98%
Bottle, 5 g
APOSPC3817B-5G
Precursor to phenethylthiazolythiourea (PETT) derivatives as potential HIV-1 reverse transcriptase inhibitors see: J. Med. Chem., 39, 4261, (1996).
- :74457-86-6
- :C₉H₉FO₂
- :MFCD00042290
- :168.17
- :80…85 °C/0,1mmHg
- :51…53 °C
- :>110 °C
- :Stue temperatur
- Pakning:Bottle
- Pakn:5 g