Order Entry
Denmark
Orders LinkContactUsLinkComponent
2'-Fluor-4'-methoxyacetophenon 98%

2'-Fluor-4'-methoxyacetophenon 98%

Bottle, 25 g
Katalog # APOSPC3817B-25G
Irriterende
Leverandør:   Apollo Scientific
2'-Fluor-4'-methoxyacetophenon 98%
Katalog # APOSPC3817B-25G
Leverandør:  Apollo Scientific
CAS nummer:  74457-86-6
2'-Fluor-4'-methoxyacetophenon 98%

Bottle, 25 g

CAS nummer:  74457-86-6
Price
220,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Precursor to phenethylthiazolythiourea (PETT) derivatives as potential HIV-1 reverse transcriptase inhibitors see: J. Med. Chem., 39, 4261, (1996).

  • CAS nummer:74457-86-6
  • Formel:C₉H₉FO₂
  • MDL nummer:MFCD00042290
  • Molecular Weight:168.17
  • Kogende Pt:80…85 °C/0,1mmHg
  • Smeltende Pt:51…53 °C
  • Flash Pt:>110 °C
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    25 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.