Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

2'-Fluor-4'-methoxyacetophenon 98%

Bottle, 100 g
Katalog # APOSPC3817B-100G
Irriterende
Leverandør:   Apollo Scientific
2'-Fluor-4'-methoxyacetophenon 98%
Katalog # APOSPC3817B-100G
Leverandør:  Apollo Scientific
CAS nummer:  74457-86-6
2'-Fluor-4'-methoxyacetophenon 98%

Bottle, 100 g

CAS nummer:  74457-86-6
Price
856,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Precursor to phenethylthiazolythiourea (PETT) derivatives as potential HIV-1 reverse transcriptase inhibitors see: J. Med. Chem., 39, 4261, (1996).

  • CAS nummer:74457-86-6
  • Formel:C₉H₉FO₂
  • MDL nummer:MFCD00042290
  • Molecular Weight:168.17
  • Kogende Pt:80…85 °C/0,1mmHg
  • Smeltende Pt:51…53 °C
  • Flash Pt:>110 °C
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    100 g

Specifikationer