Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

3,5-Dihydroxybenzonitril 98%

Bottle, 25 g
Catalog # APOSOR5346-25G
Supplier:   Apollo Scientific
3,5-Dihydroxybenzonitril 98%
Catalog # APOSOR5346-25G
Supplier:  Apollo Scientific
CAS Number:  19179-36-3
3,5-Dihydroxybenzonitril 98%

Bottle, 25 g

CAS Number:  19179-36-3
Price
2.960,00 kr
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:19179-36-3
  • Formula:C₇H₅NO₂
  • MDL Number:MFCD00016453
  • Molecular Weight:135.12 g/mol
  • Melting Pt:185…190 °C
  • Storage Temperature:Stue temperatur
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    25 g

Specifications