Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

3,5-Dihydroxybenzonitril 98%

Bottle, 1 g
Catalog # APOSOR5346-1G
Supplier:   Apollo Scientific
3,5-Dihydroxybenzonitril 98%
Catalog # APOSOR5346-1G
Supplier:  Apollo Scientific
CAS Number:  19179-36-3
3,5-Dihydroxybenzonitril 98%

Bottle, 1 g

CAS Number:  19179-36-3
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:19179-36-3
  • Formula:C₇H₅NO₂
  • MDL Number:MFCD00016453
  • Molecular Weight:135.12 g/mol
  • Melting Pt:185…190 °C
  • Storage Temperature:Stue temperatur
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.