Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
p-Cresol 99%

p-Cresol 99%

Bottle, 1 kg
Katalog # APOSOR6012-1KG
Ætsende
Giftig
Leverandør:   Apollo Scientific
p-Cresol 99%
Katalog # APOSOR6012-1KG
Leverandør:  Apollo Scientific
CAS nummer:  106-44-5
p-Cresol 99%

Bottle, 1 kg

CAS nummer:  106-44-5
Price
659,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  p-Methylphenol
  • CAS nummer:106-44-5
  • Formel:C₇H₈O
  • MDL nummer:MFCD00002376
  • Molecular Weight:108.14 g/mol
  • Kogende Pt:202 °C
  • Smeltende Pt:32…34 °C
  • Tæthed:1,034 g/cm³ (25 °C)
  • Flash Pt:85 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-398-6
  • FN:3455
  • ADR:6.1+8,II

Documents

Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    1 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.