Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
U0126 ≥99%

U0126 ≥99%

Bottle, 5 mg
Katalog # APOSBIU1095-5MG
Irriterende
Leverandør:   Apollo Scientific
U0126 ≥99%
Katalog # APOSBIU1095-5MG
Leverandør:  Apollo Scientific
CAS nummer:  109511-58-2
U0126 ≥99%

Bottle, 5 mg

CAS nummer:  109511-58-2
Price
1.380,00 kr
Log ind for tilgængelighed
1 * 5 mg
Product Details & Documents

Selective inhibitor of the mitogen-activated protein kinase kinases, MEK-1 and MEK-2, with 100-fold higher potency than PD 98059, but is, at most, a weak inhibitor of PKC, Raf, ERK, JNK, MEKK, MKK-3, MKK-4/SEK, MKK-6, Abl, Cdk2 and Cdk4. U0126 is an inhibitor of AP-1 transactivation in cell-based reporter assays. J. Biol. Chem. 273: 18623-18632 (1998).

  • CAS nummer:109511-58-2
  • Formel:C₁₈H₁₆N₆S₂
  • MDL nummer:MFCD01861186
  • Molecular Weight:380.49 g/mol
  • Smeltende Pt:172…173 °C
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    5 mg

Specifikationer