Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
U0126 ≥99%

U0126 ≥99%

Bottle, 10 mg
Katalog # APOSBIU1095-10MG
Irriterende
Leverandør:   Apollo Scientific
U0126 ≥99%
Katalog # APOSBIU1095-10MG
Leverandør:  Apollo Scientific
CAS nummer:  109511-58-2
U0126 ≥99%

Bottle, 10 mg

CAS nummer:  109511-58-2
Price
2.200,00 kr
Log ind for tilgængelighed
1 * 10 mg
Product Details & Documents

Selective inhibitor of the mitogen-activated protein kinase kinases, MEK-1 and MEK-2, with 100-fold higher potency than PD 98059, but is, at most, a weak inhibitor of PKC, Raf, ERK, JNK, MEKK, MKK-3, MKK-4/SEK, MKK-6, Abl, Cdk2 and Cdk4. U0126 is an inhibitor of AP-1 transactivation in cell-based reporter assays. J. Biol. Chem. 273: 18623-18632 (1998).

  • CAS nummer:109511-58-2
  • Formel:C₁₈H₁₆N₆S₂
  • MDL nummer:MFCD01861186
  • Molecular Weight:380.49 g/mol
  • Smeltende Pt:172…173 °C
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    10 mg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.