Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human recombinant GH Pit-20K

Human recombinant GH Pit-20K

1 mg
Katalog # APOSBITP1068-1MG
Leverandør:   Apollo Scientific
Human recombinant GH Pit-20K
Katalog # APOSBITP1068-1MG
Leverandør:  Apollo Scientific
CAS nummer:  
Human recombinant GH Pit-20K

1 mg

CAS nummer:  
Price
27.130,00 kr
Log ind for tilgængelighed
1 * 1 mg
Product Details & Documents
Specifikationer

Specifikationer

  • Pakn:
    1 mg
  • Cat. No.:
    APOSBITP1068-1MG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    GH Pit-20K
  • Protein synonyms:
    Pituitary Growth Hormone-20K
  • Species:
    Human

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.