Orders LinkContactUsLinkComponent
Benzaldehyde 99+%
Benzaldehyde 99+%
Bottle
Katalog # APOSOR45085-250G
Irriterende
Leverandør:   Apollo Scientific
Benzaldehyde 99+%
Katalog # APOSOR45085-250G
Leverandør:  Apollo Scientific
CAS nummer:  100-52-7
Benzaldehyde 99+%
Bottle
CAS nummer:  100-52-7
Price
659,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
Usage: Organic building block/ reagent.
  • CAS nummer:100-52-7
  • Formel:C₇H₆O
  • MDL nummer:MFCD00003299
  • Molecular Weight:106,12194 g/mol
  • EINECS:202-860-4
  • FN:1990
Specifikationer
Specifikationer
  • Pakning:
    Bottle
Specifikationer