Leptomycin B
Bottle, 1 mg
Katalog # APOSBIL2101-1MG
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
Potent inhibitor of nuclear export.
Supplied as a 1mM solution in abs. ethanol
Synonymer: (2E,5S,6R,7S,9R,10E,12E,15R,16Z,18E)-17-Ethyl-6-hydroxy-3,5,7,9,11,15-hexamethyl-19-((2S,3S)-3-methyl-6-oxo-3,6-dihydro-2H-pyran-2-yl)-8-oxononadeca-2,10,12,16,18-pentaenoic acid
- CAS nummer:87081-35-4
- Formel:C₃₃H₄₈O₆
- MDL nummer:MFCD06795848
- Molecular Weight:540.73 g/mol
- Flash Pt:11,1 °C
- Opbevaringstemperatur:Fryser
- FN:1170
- ADR:3,II
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:1 mg



