Orders LinkContactUsLinkComponent
Folic acid ≥98%
Folic acid ≥98%
Bottle, 25 g
Katalog # APOSBIF0608-25G
Irriterende
Leverandør:   Apollo Scientific
Folic acid ≥98%
Katalog # APOSBIF0608-25G
Leverandør:  Apollo Scientific
CAS nummer:  59-30-3
Folic acid ≥98%
Bottle, 25 g
CAS nummer:  59-30-3
Price
220,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
  • CAS nummer:59-30-3
  • Formel:C₁₉H₁₉N₇O₆
  • MDL nummer:MFCD00079305
  • Molecular Weight:441.41 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-419-0
Documents
Specifikationer
Specifikationer
  • Pakning:
    Bottle
  • Pakn:
    25 g
Specifikationer