EDTA ≥99%
Bottle, 1 kg
Katalog # APOSBIE0729-1KG
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
Synonymer: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
- CAS nummer:60-00-4
- Formel:C₁₀H₁₆N₂O₈
- MDL nummer:MFCD00003541
- Molecular Weight:292.25 g/mol
- Smeltende Pt:245 °C (dec.)
- Tæthed:1,46 g/cm³ (25 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-449-4
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:1 kg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



