Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
5,5'-Dithiobis(2-nitrobenzosyre)

5,5'-Dithiobis(2-nitrobenzosyre)

Bottle, 25 g
Katalog # APOSBID2180-25G
Irriterende
Leverandør:   Apollo Scientific
5,5'-Dithiobis(2-nitrobenzosyre)
Katalog # APOSBID2180-25G
Leverandør:  Apollo Scientific
CAS nummer:  69-78-3
5,5'-Dithiobis(2-nitrobenzosyre)

Bottle, 25 g

CAS nummer:  69-78-3
Price
933,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Used for measurement of sulphhydryl content inpeptides, proteins and tissue.

Synonymer:  DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
  • CAS nummer:69-78-3
  • Formel:C₁₄H₈N₂O₈S₂
  • MDL nummer:MFCD00007140
  • Molecular Weight:396.36 g/mol
  • Smeltende Pt:233…243 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-714-4
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.