Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
5,5'-Dithiobis(2-nitrobenzosyre)

5,5'-Dithiobis(2-nitrobenzosyre)

Bottle, 1 g
Catalog # APOSBID2180-1G
Irritant
Supplier:   Apollo Scientific
5,5'-Dithiobis(2-nitrobenzosyre)
Catalog # APOSBID2180-1G
Supplier:  Apollo Scientific
CAS Number:  69-78-3
5,5'-Dithiobis(2-nitrobenzosyre)

Bottle, 1 g

CAS Number:  69-78-3
Price
165,00 kr
Log in for availability
1 * 1 g
Product Details & Documents

Used for measurement of sulphhydryl content inpeptides, proteins and tissue.

Synonyms:  DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
  • CAS Number:69-78-3
  • Formula:C₁₄H₈N₂O₈S₂
  • MDL Number:MFCD00007140
  • Molecular Weight:396.36 g/mol
  • Melting Pt:233…243 °C
  • Storage Temperature:Stue temperatur
  • EINECS:200-714-4
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.