Hæmatoxylin
Bottle, 10 g
Katalog # APOSBIH6002-10G
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
Synonymer: DcH-48, Hydroxybrazilin, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol, NSC 2785, Natural Black I, Hydroxybrasilin, (+)-Hematoxylin
- CAS nummer:517-28-2
- Formel:C₁₆H₁₄O₆
- MDL nummer:MFCD00078111
- Molecular Weight:302.28 g/mol
- Smeltende Pt:200 °C (dec.)
- Opbevaringstemperatur:Stue temperatur
- EINECS:208-237-3
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:10 g



