Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Guanin ≥98%

Guanin ≥98%

Bottle, 25 g
Katalog # APOSBIG5137-25G
Irriterende
Leverandør:   Apollo Scientific
Guanin ≥98%
Katalog # APOSBIG5137-25G
Leverandør:  Apollo Scientific
CAS nummer:  73-40-5
Guanin ≥98%

Bottle, 25 g

CAS nummer:  73-40-5
Price
165,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  2-Amino-1H-purin-6(7H)-one
  • CAS nummer:73-40-5
  • Formel:C₅H₅N₅O
  • MDL nummer:MFCD00071533
  • Molecular Weight:151.13 g/mol
  • Smeltende Pt:>300 °C
  • Smeltende Pt:>300 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-799-8
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.