X-β-D-cellobioside
Bottle, 250 mg
Katalog # APOSBICS0106-250MG
Leverandør: Apollo Scientific
X-β-D-cellobioside
Katalog # APOSBICS0106-250MG
Leverandør: Apollo Scientific
CAS nummer: 177966-52-8
Gå til:
Product Details & Documents
Synonymer: 5-Bromo-4-chloro-3-indolyl-β-D-cellobioside
- CAS nummer:177966-52-8
- Formel:C₂₀H₂₅BrClNO₁₁
- Molecular Weight:570.78 g/mol
- Opbevaringstemperatur:Fryser
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:250 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



