Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
X-β-D-cellobioside

X-β-D-cellobioside

Bottle, 1 g
Katalog # APOSBICS0106-1G
Irriterende
Leverandør:   Apollo Scientific
X-β-D-cellobioside
Katalog # APOSBICS0106-1G
Leverandør:  Apollo Scientific
CAS nummer:  177966-52-8
X-β-D-cellobioside

Bottle, 1 g

CAS nummer:  177966-52-8
Price
483,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
Synonymer:  5-Bromo-4-chloro-3-indolyl-β-D-cellobioside
  • CAS nummer:177966-52-8
  • Formel:C₂₀H₂₅BrClNO₁₁
  • Molecular Weight:570.78 g/mol
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    1 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.