Orders LinkContactUsLinkComponent
Coconut powder
Coconut powder
Coconut powder
Katalog # APOSBIPC187-1KG
Leverandør:   Apollo Scientific
Coconut powder
Katalog # APOSBIPC187-1KG
Leverandør:  Apollo Scientific
Coconut powder
Coconut powder
Price
kr1,370.00
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
A mixture of natural coconut puree and maltodextrin. Use at a concentration of 15 to 30 g/L.
Specifikationer
  • Beskrivelse:
    Coconut powder
  • Seq:
    03