Orders LinkContactUsLinkComponent
Coconut powder
Coconut powder
Katalog # APOSBIPC187-100G
Leverandør:   Apollo Scientific
Coconut powder
Katalog # APOSBIPC187-100G
Leverandør:  Apollo Scientific
Coconut powder
Coconut powder
Price
220,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
A mixture of natural coconut puree and maltodextrin. Use at a concentration of 15 to 30 g/L.
Specifikationer
  • Beskrivelse:
    Coconut powder
  • Seq:
    01