Coconut powder
Coconut powder
Katalog # APOSBIPC187-100G
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
A mixture of natural coconut puree and maltodextrin. Use at a concentration of 15 to 30 g/L.
Specifikationer
- Beskrivelse:Coconut powder
- Seq:01