Glycin ≥99%
Bottle, 5 kg
Catalog # APOSBIG0709-5KG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Component of tris-glycine-sds running buffers forpolyacrylamide gel electrophoresis.
- Ph. Eur.
- Pyrogen free
Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS Number:56-40-6
- Formula:NH₂CH₂COOH
- MDL Number:MFCD00008131
- Molecular Weight:75.07 g/mol
- Melting Pt:240…245 °C (dec.)
- Density:1,595 g/cm³ (25 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-272-2
Specifications
Specifications
- Pakning:Bottle
- Pakn:5 kg



