(2-Hydroxyethyl)trimethylammoniumchlorid
Bottle, 500 g
Catalog # APOSBIC0605-500G
Supplier: Apollo Scientific
(2-Hydroxyethyl)trimethylammoniumchlorid
Catalog # APOSBIC0605-500G
Supplier: Apollo Scientific
CAS Number: 67-48-1
Jump to:
Product Details & Documents
Synonyms: 2-hydroxy-N,N,N-trimethylethanaminium chloride
- CAS Number:67-48-1
- Formula:HOH₂CCH₂N(CH₃)₃Cl
- MDL Number:MFCD00011721
- Molecular Weight:139.62 g/mol
- Melting Pt:302…305 °C (dec.)
- Storage Temperature:Stue temperatur
- EINECS:200-655-4
Specifications
Specifications
- Pakning:Bottle
- Pakn:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



