Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

(2-Hydroxyethyl)trimethylammoniumchlorid

Bottle, 1 kg
Katalog # APOSBIC0605-1KG
Irriterende
Leverandør:   Apollo Scientific
(2-Hydroxyethyl)trimethylammoniumchlorid
Katalog # APOSBIC0605-1KG
Leverandør:  Apollo Scientific
CAS nummer:  67-48-1
(2-Hydroxyethyl)trimethylammoniumchlorid

Bottle, 1 kg

CAS nummer:  67-48-1
Price
549,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  2-hydroxy-N,N,N-trimethylethanaminium chloride
  • CAS nummer:67-48-1
  • Formel:HOH₂CCH₂N(CH₃)₃Cl
  • MDL nummer:MFCD00011721
  • Molecular Weight:139.62 g/mol
  • Smeltende Pt:302…305 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-655-4
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    1 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.