Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

(2-Hydroxyethyl)trimethylammoniumchlorid

Bottle, 100 g
Catalog # APOSBIC0605-100G
Irritant
Supplier:   Apollo Scientific
(2-Hydroxyethyl)trimethylammoniumchlorid
Catalog # APOSBIC0605-100G
Supplier:  Apollo Scientific
CAS Number:  67-48-1
(2-Hydroxyethyl)trimethylammoniumchlorid

Bottle, 100 g

CAS Number:  67-48-1
Price
198,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2-hydroxy-N,N,N-trimethylethanaminium chloride
  • CAS Number:67-48-1
  • Formula:HOH₂CCH₂N(CH₃)₃Cl
  • MDL Number:MFCD00011721
  • Molecular Weight:139.62 g/mol
  • Melting Pt:302…305 °C (dec.)
  • Storage Temperature:Stue temperatur
  • EINECS:200-655-4
Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    100 g

Specifications