Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

2-(4-Fluorophenyl)propanoic acid

Bottle, 1 g
Katalog # APOSPC200089-1G
Giftig
Leverandør:   Apollo Scientific
2-(4-Fluorophenyl)propanoic acid
Katalog # APOSPC200089-1G
Leverandør:  Apollo Scientific
CAS nummer:  75908-73-5
2-(4-Fluorophenyl)propanoic acid

Bottle, 1 g

CAS nummer:  75908-73-5
Price
297,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
  • CAS nummer:75908-73-5
  • Formel:C₉H₉FO₂
  • MDL nummer:MFCD00054492
  • Molecular Weight:168.1650 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:278-340-6
  • FN:2811
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    1 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.