Order Entry
Denmark
Orders LinkContactUsLinkComponent
Acetohydroxamic acid

Acetohydroxamic acid

Bottle, 5 g
Katalog # APOSBIA1030-5G
Sundhed
Leverandør:   Apollo Scientific
Acetohydroxamic acid
Katalog # APOSBIA1030-5G
Leverandør:  Apollo Scientific
CAS nummer:  546-88-3
Acetohydroxamic acid

Bottle, 5 g

CAS nummer:  546-88-3
Price
165,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  N-Hydroxyacetamide
  • CAS nummer:546-88-3
  • Formel:C₂H₅NO₂
  • MDL nummer:MFCD00009994
  • Molecular Weight:75.0666 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    5 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.