Brefeldin A
Bottle, 25 mg
Katalog # APOSBIB3042-25MG
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
BFA is a macrolide antiobiotic. Disrupts mechanism of Golgi apparatus.
Synonymer: (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural
- CAS nummer:20350-15-6
- Formel:C₁₆H₂₄O₄
- MDL nummer:MFCD00083258
- Molecular Weight:280.4 g/mol
- Smeltende Pt:200…205 °C
- Opbevaringstemperatur:Køleskab
- FN:2811
- ADR:6.1,III
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:25 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



