Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
MES monohydrat

MES monohydrat

Bottle, 250 g
Catalog # APOSBIB2012-250G
Irritant
Supplier:   Apollo Scientific
MES monohydrat
Catalog # APOSBIB2012-250G
Supplier:  Apollo Scientific
CAS Number:  145224-94-8
MES monohydrat

Bottle, 250 g

CAS Number:  145224-94-8
Price
878,00 kr
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
  • CAS Number:145224-94-8
  • Formula:C₆H₁₃NO₄S·1H₂O
  • MDL Number:MFCD00149409
  • Molecular Weight:213.25 g/mol
  • Melting Pt:308 °C
  • Storage Temperature:Køleskab
  • EINECS:224-632-3

Documents

Specifications

Specifications

  • Pakning:
    Bottle
  • Pakn:
    250 g

Specifications