Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L-Alanin

L-Alanin

Bottle, 100 g
Katalog # APOSBIA0703-100G
Irriterende
Leverandør:   Apollo Scientific
L-Alanin
Katalog # APOSBIA0703-100G
Leverandør:  Apollo Scientific
CAS nummer:  56-41-7
L-Alanin

Bottle, 100 g

CAS nummer:  56-41-7
Price
220,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

USP, pyrogen free

Synonymer:  (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
  • CAS nummer:56-41-7
  • Formel:C₃H₇NO₂
  • MDL nummer:MFCD00064410
  • Molecular Weight:89.09 g/mol
  • Smeltende Pt:314…316 °C (dec.)
  • Tæthed:1,432 g/cm³ (25 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-273-8

Documents

Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.