ACES ≥99%
Bottle, 500 g
Katalog # APOSBIA0301-500G
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
An efficient separator in the resolution of proteinsystems by IEF. Biological buffer - useful range 6.4 - 7.4
Synonymer: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid
- CAS nummer:7365-82-4
- Formel:H₂NCOCH₂NHCH₂CH₂SO₃H
- MDL nummer:MFCD00008030
- Molecular Weight:182.2 g/mol
- Smeltende Pt:270…295 °C (dec.)
- Opbevaringstemperatur:Stue temperatur
- EINECS:230-908-4
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:500 g



