ACES ≥99%
Bottle, 250 g
Katalog # APOSBIA0301-250G
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
An efficient separator in the resolution of proteinsystems by IEF. Biological buffer - useful range 6.4 - 7.4
Synonymer: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid
- CAS nummer:7365-82-4
- Formel:H₂NCOCH₂NHCH₂CH₂SO₃H
- MDL nummer:MFCD00008030
- Molecular Weight:182.2 g/mol
- Smeltende Pt:270…295 °C (dec.)
- Opbevaringstemperatur:Stue temperatur
- EINECS:230-908-4
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:250 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



