Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

ACES ≥99%

Bottle, 100 g
Katalog # APOSBIA0301-100G
Irriterende
Leverandør:   Apollo Scientific
ACES ≥99%
Katalog # APOSBIA0301-100G
Leverandør:  Apollo Scientific
CAS nummer:  7365-82-4
ACES ≥99%

Bottle, 100 g

CAS nummer:  7365-82-4
Price
275,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

An efficient separator in the resolution of proteinsystems by IEF. Biological buffer - useful range 6.4 - 7.4

Synonymer:  ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid
  • CAS nummer:7365-82-4
  • Formel:H₂NCOCH₂NHCH₂CH₂SO₃H
  • MDL nummer:MFCD00008030
  • Molecular Weight:182.2 g/mol
  • Smeltende Pt:270…295 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:230-908-4
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.