Natriumpyruvat
Bottle, 500 g
Katalog # APOSBIB6270-500G
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
Medium of animal cell culture; Material for pharmaceuticals, agrochemicals and diagnostic reagent; Intermediate for flavors and fragrances.
Synonymer: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS nummer:113-24-6
- Formel:CH₃COCOONa
- MDL nummer:MFCD00002586
- Molecular Weight:110.04 g/mol
- Smeltende Pt:>300 °C
- Smeltende Pt:>300 °C
- Opbevaringstemperatur:Køleskab
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:500 g



