Ethidiumbromid ≥98% for biotechnology
250 g
Leverandør: VWR Chemicals
Ethidiumbromid ≥98% for biotechnology
Katalog # 0492-CHR-250G
Leverandør: VWR Chemicals
CAS nummer: 1239-45-8
Gå til:
Product Details & Documents
Fluorescent stain for visualizing nucleic acids.
At a concentration of 0.5 μg/ml, ethidium bromide is useful in agarose or acrylamide gels for band detection. It is used to detect as little as 10 ng of nucleic acid. At higher concentrations (0.5 to 1.0 μg/ml), it may be used to facilitate purification of DNA in a cesium chloride gradient.
For maximum convenience, VWR offers ethidium bromide in various forms, including ready-to-use solutions and powdered forms. Each is packaged in shatter-resistant packaging for added safety. VWR’s ethidium bromide dropper bottle offers the ultimate convenience and safety. Simply dispense one drop of ethidium bromide into 50ml of agarose gel or running buffer (TBE or TAE) or staining solution.
Synonymer: 3,8-Diamino-5-ethyl-6-phenylphenantridiniumbromid
- CAS nummer:1239-45-8
- Formel:C₂₁H₂₀BrN₃
- MDL nummer:MFCD00011724
- Molecular Weight:394.31 g/mol
- Merck Index:13,04751
- Kogende Pt:72 °C (0,1 Torr)
- Smeltende Pt:261…264 °C
- Tæthed:1,01 g/cm³ (20 °C)
- Flash Pt:<100 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:214-984-6
- FN:2811
- ADR:6.1,I
Specifikationer
Specifikationstestresultater
- DNase:None Detected
- Loss on Drying:≤5%
- Protease:None Detected
- Purity (Dry Basis):≥98%
- RNase:None
Specifikationer
- Pakn:250 g



