Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Optimum Growth Flask

Optimum Growth™ flask, 250 ml
Katalog # THMS931111
Leverandør:   THOMSON INSTRUMENT
Optimum Growth Flask
Katalog # THMS931111
Leverandør:  THOMSON INSTRUMENT
Optimum Growth Flask
Optimum Growth Flask

Optimum Growth™ flask, 250 ml

Price
5.770,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents

Optimum Growth™ flasks (patented) give excellent growth and expression for both mammalian and insect cell lines.

  • Baffles designed for high aeration and low shear
  • Same footprint as compairable Fernbach flask
  • Less foaming than disposable Fernbach flask
  • Transfer cap connects directly to Wave Bags™ and bioreactors with quick connect or tube fusing
  • 0,2 μm vented cap
  • Individually packaged and sterilised

 

Specifikationer
  • Kapacitet:
    250 ml
  • Beskrivelse:
    Optimum Growth™ flask
  • Seq:
    2