DCpak® SFC Achiral (SFC- B) Columns
Analytisk, DCpak SFC-B, 3 µm
Katalog # DAICABS92
Leverandør: CHIRAL TECHNOLOGIES
Gå til:
Product Details & Documents
When a material is in a state that the temperature and the pressure exceed the critical point, it becomes a supercritical fluid. In this phase, the density of gas and liquid becomes the same.
Specifikationer
- Kolonne form:Analytisk
- Partikelstørrelse:3 µm
- Seq:03
- Fase:DCpak SFC-B