High-Performance Pharmacy Refrigerator, TSX
Product Details & Documents
Thermo Scientific™ TSX Series high-performance pharmacy refrigerators are designed with features that support sample protection and sustainability objectives for the storage of pharmaceuticals, vaccines, chemotherapy and other medical and pharmacy-grade storage requiring 2 to 8 °C. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection. Certified to NSF/ANSI 456 Vaccine Storage standard, the TSX series can help you meet regulations and guidelines from the CDC, World Health Organisation, and other Health Ministries on vaccine storage and handling. The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
- Triple panel glass door helps maintain optimal internal conditions
- Forced-air circulation for temperature uniformity and fast temperature recovery
- Heat-free defrost for maximum temperature uniformity
- Quiet operation at just 52 dBA
- Keyed on/off switch with easy to use, setpoint security
- Self-closing door with 90° stop to assist with inventory loads
- Built-in remote alarm contacts
V-drive technology: While conventional refrigerant, high-performance refrigerators and freezers use single speed compressors that continually cycle on and off, the TSX Series V-drive runs at variable speeds to adjust cooling performance to the conditions inside and outside of the refrigerator. When combined with the automated tuning control, this variable speed drive optimizes the compressors running speed to the current conditions. When there are frequent door openings, or samples are added to the refrigerator, the control system detects the activity and increases the drive speed to bring temperatures back to the set point quickly. This innovative technology is one of the reasons the TSX Series delivers outstanding door opening recovery (DOR) speed, and more peace of mind for busy laboratories. When conditions are stable, such as when the unit is running overnight or on weekends, the drive runs at a low speed, reducing energy consumption while maintaining a stable temperature for the samples.
Tested to GMP Clean Room Class A/ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP and hazardous drug compounding and other clean room environments. Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863. Ergonomically designed door handle with a soft-touch grip. The microprocessor controller is angled to help prevent neck strain, Convenient, fully extendable and adjustable basket drawers, Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction. Self-closing door with 90° stop to assist with inventory loads, US EPA ENERGY STAR Certified with up to 40% less energy usage than conventional refrigerant models. ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations. Manufactured in an award-winning, zero-waste to landfill facility. Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always and anywhere know the safety of your samples and the performance of your refrigerator or freezer. Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems. Alternate door swing configuration and door types. Extra and alternate wire baskets, shelves, and drawers. Free-standing or factory installed chart recorders. Stainless steel interior finish.
E-Lock adapter kits, 4 to 20 milliamp transmitters and other remote alarm modules. Specialty, regional, and country specific plugs. Industry Leading 120-month compressor warranty (varies by country, please contact us for details). Included Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate. Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs, Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs.
: UL, cUL, CE.
Specifikationer
- B×D×H udv.:711×940×1996 mm
- Kapacitet:650 L
- Beskrivelse:Pharmacy refrigerator, TSX2305PV
- Seq:02
- B×D×H indv.:61×723×1473 mm
- Temperatur:2...8 °C
- Stik:CEE 7/7
- Art.nr:HERATSX2305PV
- Environmentally Preferable:Energieffektiv|Sikrere sundhed for mennesker og miljø
- Dørtype:1 glass swing door
- Shipping weight:231 kg
- Cat. No.:HERATSX2305PV
- Specifikationer:4 casters; the front 2 are lockable
Bright, LED interior lighting
230 V, 16 A, 50 Hz - Kølemiddel:R290
- Hylder/skuffer:6 fully extendable and adjustable wire baskets
- Alarm:Audible and visual
- Vægt:237 kg
- Model:TSX2305PV
- Elektriske specifikationer:208-230 V, 50 Hz
- Afrimning:Automatic
- Display:Microprocessor
Services
Produktinformation
- Katalog #BeskrivelseTilgængelighedPrice
- SERW1100.0900.01Forebyggende vedligeholdelse og driftstest af køleskabet on-site. Reservedele og rejseudgifter er ikke inkluderet.Log ind for tilgængelighed1 * 1 Each - 2.060,00 kr
- SERW1100.7000.01Installationskvalificering og driftskvalificering af køleskab on-site. Kvalifikationsprotokoller er ikke inkluderet. Rejseudgifter er inkluderet (ekskl. udgifter til flyrejser).Log ind for tilgængelighed1 * 1 Each - 42.950,00 kr
- SERW1100.1101.01Forebyggende vedligeholdelse, driftsprøvning og kalibrering af køleskabet on-site. 1 Temperatur (1 probe) Kalibrering . Kalibreringscertifikat medfølger. Reservedele og rejseudgifter ikke inkluderet.Log ind for tilgængelighed1 * 1 Each - 3.090,00 kr
- SERW1100.0100.01Installation af køleskab on-site. Enheden skal være på anvendelsesstedet. Funktionstest og kort introduktion af brugernes korte funktioner. Rejseudgifter inkluderet (ekskl. udgifter til flyrejser).Log ind for tilgængelighed1 * 1 Each - 12.280,00 kr



