Orders LinkContactUsLinkComponent
Cleaning coupons
Cleaning coupons
Cleaning coupon, 316 stainless steel (bright polish), 100×100 mm
Katalog # SAMP8644S6-100B
Leverandør:   Sampling Systems
Cleaning coupons
Katalog # SAMP8644S6-100B
Leverandør:  Sampling Systems
Cleaning coupons
Cleaning coupons
Cleaning coupon, 316 stainless steel (bright polish), 100×100 mm
Price
2.540,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These cleaning validation coupons are available in all popular materials and various sizes.
Specifikationer
  • Beskrivelse:
    Cleaning coupon, 316 stainless steel (bright polish)
  • L×B:
    100×100 mm
  • Seq:
    04