Qualitix® Pipette Tips
Tipfill system, incl. ultra-micro tips, fine point, 10 µl, -, Transparent, Tipfill system (13 racks, 96 each)
Katalog # SOCO3090.0010T
Leverandør: Socorex Isba
Gå til:
Product Details & Documents
These tips are made of 100% pure virgin, medical grade PP, fitting not only Socorex® but all major micropipette brands.
- Superior quality, autoclavable
- Lot by lot consistency
- Certified free of nuclease, endotoxin and ATP
- Reduced packaging size, less bench space requirement
- Broad compatibility with other micropipette brands
Beveled tips: Special shape eliminates liquid drop sticking at tip end.
Fine pointed tips: Have a thin, soft end enabling smooth liquid deposit.
Low binding tips are made with a silicone-free resin reducing surface adhesion. They provide maximal liquid recovery. Best suited for proteomic research and handling of any viscous liquids. Protease-free certified.
: The tipfill system is intended for easy rack loading. Tip trays are instantly transferred into a rack without risk of contamination. The smart re-closable tipfill system box reduces storage space, waste volume and shipping costs.
Specifikationer
- Pakket:Tipfill system (13 racks, 96 each)
- Steril:-
- Volumen:10 µl
- Spidstype:Tipfill system, incl. ultra-micro tips, fine point
- Spits farge:Transparent
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Empty rack for 10/200 µl tips, to be used with tipfill systemEmpty rack for 10/200 µl tips, to be used with tipfill systemSOCO3070.0210ERLog ind for tilgængelighed1 * 10 Items - 268,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Empty rack for 1000 µl tips, to be used with tipfill systemEmpty rack for 1000 µl tips, to be used with tipfill systemSOCO3070.1000ERLog ind for tilgængelighed1 * 6 Items - 230,00 kr