Orders LinkContactUsLinkComponent
Empty racks for tipfill system, Qualitix®
Empty racks for tipfill system, Qualitix®
Empty rack for 1000 µl tips, to be used with tipfill system
Katalog # SOCO3070.1000ER
Leverandør:   Socorex Isba
Empty racks for tipfill system, Qualitix®
Katalog # SOCO3070.1000ER
Leverandør:  Socorex Isba
Empty racks for tipfill system, Qualitix®
Empty racks for tipfill system, Qualitix®
Empty rack for 1000 µl tips, to be used with tipfill system
Price
230,00 kr
Log ind for tilgængelighed
1 * 6 Items
Product Details & Documents
These empty racks are designed to be used with Qualitix® tipfill packs.
  • Racks have smooth rounded edges and ergonomic snap latch that won’t catch gloves
  • Transparent lid enables instant view of content
  • Empty racks are re-usable and autoclavable
Specifikationer
  • Beskrivelse:
    Empty rack for 1000 µl tips, to be used with tipfill system
  • Seq:
    02