Magnetic stirring bars
Magnetic stirring bar Ø 6×35 mm, 35 mm, 6 mm, AREC series
Katalog # VELPA00001056
Leverandør: VELP SCIENTIFIC
Gå til:
Product Details & Documents
Specifikationer
- Ø:6 mm
- Til:AREC series
- Beskrivelse:Magnetic stirring bar Ø 6×35 mm
- Længde:35 mm
- Seq:05