Magnetic stirring bars
Magnetic stirring bar Ø 8×40 mm, 40 mm, 8 mm, AREC series
Katalog # VELPA00000356
Leverandør: VELP SCIENTIFIC
Gå til:
Product Details & Documents
Specifikationer
- Ø:8 mm
- Til:AREC series
- Beskrivelse:Magnetic stirring bar Ø 8×40 mm
- Længde:40 mm
- Seq:08