Orders LinkContactUsLinkComponent
Magnetic stirring bars
Magnetic stirring bars
Magnetic stirring bar, 10 mm, 5 mm
Katalog # VELPA00000336
Leverandør:   VELP SCIENTIFIC
Magnetic stirring bars
Katalog # VELPA00000336
Leverandør:  VELP SCIENTIFIC
Magnetic stirring bars
Magnetic stirring bars
Magnetic stirring bar, 10 mm, 5 mm
Price
30,80 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Ø:
    5 mm
  • Beskrivelse:
    Magnetic stirring bar
  • Længde:
    10 mm
  • Seq:
    03